SY3H3D680477 Synthonix Corporation

4-Methylpyrazolo[1,5-a]pyridine-3-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 143803-93-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link https://synthonix.com/51402.html

Safety Information

MSDS https://synthonix.com/51402.html
Availability details for
Quantity
  • Availability
  • Add to Cart