SY3H3D68047C Synthonix Corporation

Methyl 3-aminothieno[3,2-c]pyridine-2-carboxylate

  • MDL number No MDL found
  • Cas Number : 111042-92-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/51409.html

Safety Information

MSDS https://synthonix.com/51409.html
Availability details for
Quantity
  • Availability
  • Add to Cart