SY3H3D68047D Synthonix Corporation

Methyl 2-mercapto-1H-benzo[d]imidazole-4-carboxylate

  • MDL number No MDL found
  • Cas Number : 92807-02-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/51410.html

Safety Information

MSDS https://synthonix.com/51410.html
Availability details for
Quantity
  • Availability
  • Add to Cart