SY3H3D680481 Synthonix Corporation

7-Methoxy-4-nitro-1H-indole

  • MDL number No MDL found
  • Cas Number : 175913-32-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/51414.html

Safety Information

MSDS https://synthonix.com/51414.html
Availability details for
Quantity
  • Availability
  • Add to Cart