SY3H3D680482 Synthonix Corporation

5-Methoxyquinazoline-2,4(1H,3H)-dione

  • Cas Number : 61948-86-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/51417.html

Safety Information

MSDS https://synthonix.com/51417.html
Availability details for
Quantity
  • Availability
  • Add to Cart