SY3H3D680484 Synthonix Corporation

4-Methoxypyrazolo[1,5-a]pyridine-3-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 1378695-64-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/51422.html

Safety Information

MSDS https://synthonix.com/51422.html
Availability details for
Quantity
  • Availability
  • Add to Cart