SY3H3D680485 Synthonix Corporation

5-Methoxypyrazolo[1,5-a]pyridine-3-carboxylic acid

  • Cas Number : 99446-32-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/51423.html

Safety Information

MSDS https://synthonix.com/51423.html
Availability details for
Quantity
  • Availability
  • Add to Cart