SY3H3D680488 Synthonix Corporation

6-Methylquinoxaline-2,3-dithiol

  • MDL number No MDL found
  • Cas Number : 25625-62-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/51431.html

Safety Information

MSDS https://synthonix.com/51431.html
Availability details for
Quantity
  • Availability
  • Add to Cart