SY3H3D680490 Synthonix Corporation

Methyl 3-(bromomethyl)-2-nitrobenzoate

  • MDL number No MDL found
  • Cas Number : 132874-06-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/51488.html

Safety Information

MSDS https://synthonix.com/51488.html
Availability details for
Quantity
  • Availability
  • Add to Cart