SY3H3D680494 Synthonix Corporation

Methyl 3-bromo-4-methyl-5-nitrobenzoate

  • MDL number No MDL found
  • Cas Number : 223519-08-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/51492.html

Safety Information

MSDS https://synthonix.com/51492.html
Availability details for
Quantity
  • Availability
  • Add to Cart