SY3H3D680497 Synthonix Corporation

Methyl 2-fluoro-5-methyl-3-nitrobenzoate

  • Cas Number : 1365988-42-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/51501.html

Safety Information

MSDS https://synthonix.com/51501.html
Availability details for
Quantity
  • Availability
  • Add to Cart