SY3H3D680498 Synthonix Corporation

Methyl 5-fluoro-2-methyl-3-nitrobenzoate

  • Cas Number : 697739-03-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/51502.html

Safety Information

MSDS https://synthonix.com/51502.html
Availability details for
Quantity
  • Availability
  • Add to Cart