SY3H3D68049A Synthonix Corporation

6-Methoxybenzofuran

  • MDL number No MDL found
  • Cas Number : 50551-63-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link https://synthonix.com/51550.html

Safety Information

MSDS https://synthonix.com/51550.html
Availability details for
Quantity
  • Availability
  • Add to Cart