SY3H3D68049C Synthonix Corporation

5-Methylisobenzofuran-1(3H)-one

  • MDL number No MDL found
  • Cas Number : 54120-64-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/51553.html

Safety Information

MSDS https://synthonix.com/51553.html
Availability details for
Quantity
  • Availability
  • Add to Cart