SY3H3D68049D Synthonix Corporation

6-Methylisobenzofuran-1(3H)-one

  • MDL number No MDL found
  • Cas Number : 72985-23-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/51554.html

Safety Information

MSDS https://synthonix.com/51554.html
Availability details for
Quantity
  • Availability
  • Add to Cart