SY3H3D6804A0 Synthonix Corporation

Methyl 4-bromo-3-(bromomethyl)benzoate

  • MDL number No MDL found
  • Cas Number : 142031-67-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/51564.html

Safety Information

MSDS https://synthonix.com/51564.html
Availability details for
Quantity
  • Availability
  • Add to Cart