SY3H3D6804A2 Synthonix Corporation

Methyl 2-(bromomethyl)-3-chlorobenzoate

  • MDL number No MDL found
  • Cas Number : 188187-03-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/51567.html

Safety Information

MSDS https://synthonix.com/51567.html
Availability details for
Quantity
  • Availability
  • Add to Cart