SY3H3D6804A8 Synthonix Corporation

Methyl 5-(bromomethyl)-2-fluorobenzoate

  • Cas Number : 709-45-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/51582.html

Safety Information

MSDS https://synthonix.com/51582.html
Availability details for
Quantity
  • Availability
  • Add to Cart