SY3H3D6804AA Synthonix Corporation

Methyl 2-bromo-2-(4-fluorophenyl)acetate

  • MDL number MFCD08444284
  • Cas Number : 71783-54-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Pale-yellow to Yellow-brown Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/51587.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/51587.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart