SY3H3D6804AB Synthonix Corporation

Methyl 2-bromo-4-fluoro-6-methylbenzoate

  • Cas Number : 1262396-04-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/51588.html

Safety Information

MSDS https://synthonix.com/51588.html
Availability details for
Quantity
  • Availability
  • Add to Cart