SY3H3D6804AC Synthonix Corporation

Methyl 3-bromo-5-fluoro-4-methylbenzoate

  • Cas Number : 1533932-57-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/51589.html

Safety Information

MSDS https://synthonix.com/51589.html
Availability details for
Quantity
  • Availability
  • Add to Cart