SY3H3D6804AD Synthonix Corporation

Methyl 4-bromo-2-fluoro-6-methylbenzoate

  • Cas Number : 1427409-40-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/51590.html

Safety Information

MSDS https://synthonix.com/51590.html
Availability details for
Quantity
  • Availability
  • Add to Cart