SY3H3D6804AF Synthonix Corporation

Methyl 2-(2,6-dichlorophenyl)acetate

  • MDL number No MDL found
  • Cas Number : 54551-83-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/51600.html

Safety Information

MSDS https://synthonix.com/51600.html
Availability details for
Quantity
  • Availability
  • Add to Cart