SY3H3D6804B7 Synthonix Corporation

Methyl 4-bromo-2-fluoro-6-methoxybenzoate

  • MDL number No MDL found
  • Cas Number : 1427415-25-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/51633.html

Safety Information

MSDS https://synthonix.com/51633.html
Availability details for
Quantity
  • Availability
  • Add to Cart