SY3H3D6804BE Synthonix Corporation

2-Methyl-1,4-benzenedicarboxylic acid

  • MDL number No MDL found

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/51643.html

Safety Information

MSDS https://synthonix.com/51643.html
Availability details for
Quantity
  • Availability
  • Add to Cart