SY3H3D6804C7 Synthonix Corporation

6-Methylbenzo[b]thiophene

  • MDL number No MDL found
  • Cas Number : 16587-47-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/51666.html

Safety Information

MSDS https://synthonix.com/51666.html
Availability details for
Quantity
  • Availability
  • Add to Cart