SY3H3D6804C9 Synthonix Corporation

2-(1-Methyl-1H-pyrazol-5-yl)pyridine

  • MDL number No MDL found
  • Cas Number : 938066-21-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/51707.html

Safety Information

MSDS https://synthonix.com/51707.html
Availability details for
Quantity
  • Availability
  • Add to Cart