SY3H3D6804CB Synthonix Corporation

4-(1-Methyl-1H-pyrazol-5-yl)pyridine

  • Cas Number : 905281-60-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/51709.html

Safety Information

MSDS https://synthonix.com/51709.html
Availability details for
Quantity
  • Availability
  • Add to Cart