SY3H3D6804CC Synthonix Corporation

Methyl 3-amino-1H-indazole-6-carboxylate

  • Cas Number : 1279865-95-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link https://synthonix.com/51724.html

Safety Information

MSDS https://synthonix.com/51724.html
Availability details for
Quantity
  • Availability
  • Add to Cart