SY3H3D6804CD Synthonix Corporation

Methyl 6-amino-1H-indazole-7-carboxylate

  • MDL number No MDL found
  • Cas Number : 73907-98-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/51725.html

Safety Information

MSDS https://synthonix.com/51725.html
Availability details for
Quantity
  • Availability
  • Add to Cart