SY3H3D6804CF Synthonix Corporation

4-(5-(Methylsulfonyl)-1,3,4-oxadiazol-2-yl)aniline

  • Cas Number : 1858267-88-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/51736.html

Safety Information

MSDS https://synthonix.com/51736.html
Availability details for
Quantity
  • Availability
  • Add to Cart