SY3H3D6804D2 Synthonix Corporation

N-(3-(5-Mercapto-1H-tetrazol-1-yl)phenyl)acetamide

  • Cas Number : 14070-48-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/51743.html

Safety Information

MSDS https://synthonix.com/51743.html
Availability details for
Quantity
  • Availability
  • Add to Cart