SY3H3D6804D3 Synthonix Corporation

4-Methylindolin-2-one

  • MDL number No MDL found
  • Cas Number : 13220-46-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/51751.html

Safety Information

MSDS https://synthonix.com/51751.html
Availability details for
Quantity
  • Availability
  • Add to Cart