SY3H3D6804D6 Synthonix Corporation

2-Methoxy-4-methylbenzonitrile

  • MDL number No MDL found
  • Cas Number : 53078-69-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/51760.html

Safety Information

MSDS https://synthonix.com/51760.html
Availability details for
Quantity
  • Availability
  • Add to Cart