SY3H3D6804D7 Synthonix Corporation

2-Methoxy-6-methylbenzonitrile

  • MDL number No MDL found
  • Cas Number : 53005-44-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/51761.html

Safety Information

MSDS https://synthonix.com/51761.html
Availability details for
Quantity
  • Availability
  • Add to Cart