SY3H3D6804D8 Synthonix Corporation

3-Methoxy-4-methylbenzonitrile

  • Cas Number : 3556-60-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/51762.html

Safety Information

MSDS https://synthonix.com/51762.html
Availability details for
Quantity
  • Availability
  • Add to Cart