SY3H3D6804DF Synthonix Corporation

(2-Methylbenzo[d]oxazol-5-yl)methanol

  • MDL number No MDL found
  • Cas Number : 136663-38-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/51784.html

Safety Information

MSDS https://synthonix.com/51784.html
Availability details for
Quantity
  • Availability
  • Add to Cart