SY3H3D6804E7 Synthonix Corporation

1-(4-Methyl-2-nitrophenyl)ethanone

  • MDL number No MDL found
  • Cas Number : 155694-84-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/51820.html

Safety Information

MSDS https://synthonix.com/51820.html
Availability details for
Quantity
  • Availability
  • Add to Cart