SY3H3D6804F0 Synthonix Corporation

Methyl 2-methyl-5-nitrobenzoate

  • Cas Number : 77324-87-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/51835.html

Safety Information

MSDS https://synthonix.com/51835.html
Availability details for
Quantity
  • Availability
  • Add to Cart