SY3H3D6804F1 Synthonix Corporation

Methyl 2-methyl-6-nitrobenzoate

  • MDL number No MDL found
  • Cas Number : 61940-22-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/51836.html

Safety Information

MSDS https://synthonix.com/51836.html
Availability details for
Quantity
  • Availability
  • Add to Cart