SY3H3D6804F3 Synthonix Corporation

Methyl 5-methyl-2-nitrobenzoate

  • Cas Number : 20587-30-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/51838.html

Safety Information

MSDS https://synthonix.com/51838.html
Availability details for
Quantity
  • Availability
  • Add to Cart