SY3H3D6804F6 Synthonix Corporation

2-(3-Methyl-2-nitrophenyl)acetic acid

  • MDL number No MDL found
  • Cas Number : 18710-86-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/51844.html

Safety Information

MSDS https://synthonix.com/51844.html
Availability details for
Quantity
  • Availability
  • Add to Cart