SY3H3D6804FD Synthonix Corporation

2-(3-Methoxy-4-nitrophenyl)acetic acid

  • MDL number No MDL found
  • Cas Number : 5803-22-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/51856.html

Safety Information

MSDS https://synthonix.com/51856.html
Availability details for
Quantity
  • Availability
  • Add to Cart