SY3H3D6804FF Synthonix Corporation

5-Methoxy-4-methyl-2-nitrobenzoic acid

  • MDL number No MDL found
  • Cas Number : 857599-32-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/51859.html

Safety Information

MSDS https://synthonix.com/51859.html
Availability details for
Quantity
  • Availability
  • Add to Cart