SY3H3D680508 Synthonix Corporation

Methyl 2-bromo-3-methylbenzoate

  • MDL number No MDL found
  • Cas Number : 131001-86-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/51892.html

Safety Information

MSDS https://synthonix.com/51892.html
Availability details for
Quantity
  • Availability
  • Add to Cart