SY3H3D68050E Synthonix Corporation

Methyl 4-bromo-2-methylbenzoate

  • MDL number MFCD09954957
  • Cas Number : 99548-55-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Liquid or semi-solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/51898.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/51898.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart