SY3H3D68051A Synthonix Corporation

Methyl 2-fluoro-3-methylbenzoate

  • MDL number No MDL found
  • Cas Number : 586374-04-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/51940.html

Safety Information

MSDS https://synthonix.com/51940.html
Availability details for
Quantity
  • Availability
  • Add to Cart