SY3H3D68051D Synthonix Corporation

Methyl 4-fluoro-3-methylbenzoate

  • MDL number No MDL found
  • Cas Number : 180636-50-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/51943.html

Safety Information

MSDS https://synthonix.com/51943.html
Availability details for
Quantity
  • Availability
  • Add to Cart