SY3H3D680524 Synthonix Corporation

Methyl 4-iodo-3-methylbenzoate

  • MDL number MFCD02683863
  • Cas Number : 5471-81-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: White - Pale yellow red - Yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/51963.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/51963.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart