SY3H3D680525 Synthonix Corporation

Methyl 5-iodo-2-methylbenzoate

  • MDL number No MDL found
  • Cas Number : 103440-54-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/51964.html

Safety Information

MSDS https://synthonix.com/51964.html
Availability details for
Quantity
  • Availability
  • Add to Cart